Merck
  • Reactive sulfur species regulate tRNA methylthiolation and contribute to insulin secretion.
Reactive sulfur species regulate tRNA methylthiolation and contribute to insulin secretion.
Nucleic acids research (2016-08-29)
Nozomu Takahashi, Fan-Yan Wei, Sayaka Watanabe, Mayumi Hirayama, Yuya Ohuchi, Atsushi Fujimura, Taku Kaitsuka, Isao Ishii, Tomohiro Sawa, Hideki Nakayama, Takaaki Akaike, Kazuhito Tomizawa
ABSTRACT
The 2-methylthio (ms
Sigma-Aldrich
MISSION® esiRNA, targeting human CBS
Sigma-Aldrich
DL-Cysteine, technical grade