Merck
  • AMPK-autophagy inhibition sensitizes icaritin-induced anti-colorectal cancer cell activity.
AMPK-autophagy inhibition sensitizes icaritin-induced anti-colorectal cancer cell activity.
Oncotarget (2017-01-20)
Chunxian Zhou, Jun Gu, Gang Zhang, Da Dong, Qunying Yang, Min-Bin Chen, Dongfeng Xu
ABSTRACT
The current research studied the potential effect of autophagy on icaritin-induced anti-colorectal cancer (CRC) cell activity. Treatment of icaritin in both primary and established (HT-29) CRC cells induced feedback activation of autophagy, evidenced by p62 degradation, Beclin-1 and autophagy-related gene-5 (ATG-5) upregulation, as well as light chain 3B (LC3B)-GFP puncta formation. Pharmacological inhibiting of autophagy dramatically potentiated icaritin-induced CRC cell death and apoptosis. Meanwhile, shRNA-mediated knockdown of Beclin-1 or ATG-5 also sensitized icaritin-induced CRC cell death and apoptosis. Icaritin activated AMP-activated protein kinase (AMPK) signaling in CRC cells, functioning as the upstream signaling for autophagy activation. shRNA/siRNA-mediated knockdown of AMPKα1inhibited icaritin-induced autophagy activation, but exacerbated CRC cell death. On the other hand, the AMPK activator compound 13 (C13) or the autophagy activator MHY1485 attenuated icaritin-induced cytotoxicity. In nude mice, icaritin (oral administration)-induced HT-29 tumor growth inhibition was potentiated when combined with AMPKα1 shRNA knockdown in tumors. We conclude that feedback activation of AMPK-autophagy pathway could be a primary resistance factor of icaritin.
Sigma-Aldrich
MISSION® esiRNA, targeting human BECN1
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor