Merck
Mechanosensitivity of polydiacetylene with a phosphocholine headgroup.
Soft matter (2017-02-07)
Roberto Diego Ortuso, Ugo Cataldi, Kaori Sugihara
ABSTRACT
We have demonstrated colorimetric and fluorescence detection of a peptide, melittin, based on polydiacetylene (PDA) made of 1,2-bis(10,12-tricosadiynoyl)-sn-glycero-3-phosphocholine (DC(8,9)PC). The PDA used in this work has a phosphocholine headgroup, which mimics peptide-cell membrane interactions better than the conventional PDAs with carboxyl headgroups. The dose curve (colorimetric response vs. melittin concentration) showed a half maximum response at the melittin concentration of 0.1 mg ml
Sigma-Aldrich
3-(Benzyldimethylammonio)propanesulfonate, BioXtra, ≥99.0% (HPCE)
Sigma-Aldrich
10,12-Tricosadiynoic acid, ≥98.0% (GC)