Merck
  • Effect of QiShenYiQi pill on myocardial collagen metabolism in experimental autoimmune myocarditis rats.
Effect of QiShenYiQi pill on myocardial collagen metabolism in experimental autoimmune myocarditis rats.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2017-02-09)
Shi-Chao Lv, Meifang Wu, Meng Li, Qiang Wang, Xiao-Jing Wang, Ao Zhang, Ling Xu, Jun-Ping Zhang
ABSTRACT
To observe the effect of QiShenYiQi pill (QSYQ) on myocardial collagen metabolism in experimental autoimmune myocarditis rats, and to explore its mechanism of action. Lewis rats underwent the injection of myocardial myosin mixed with freund's complete adjuvant were randomized into three groups: model, valsartan and QSYQ groups. And we treated rats which were injected phosphate buffered saline (PBS) mixed with freund's complete adjuvant as control group. Rats were intervened and euthanized at 4 and 8 weeks. We use alkaline hydrolysis to detect the content of myocardial hydroxyproline (HYP), and ELISA to detect the level of serum procollagen type I carboxyterminal peptide (PICP), procollagen type III amino-terminal peptide (PIIINP), and collagen C telopeptide type I (CTX-I). Myocardial MMP-1 and TIMP-1 protein expression was detected by immunohistochemistry, and myocardial MMP-1 and TIMP-1 mRNA expression was detected by real-time qPCR. QSYQ reduced the content of myocardial HYP, and this reduction was greater over time. QSYQ also reduced the serum concentration of PICP, PIIINP, CTX-I and the PICP/PIIINP ratio, which further reduced over time, whereas its effect on lowering PICP was significantly greater than that of valsartan at 4 and 8 weeks, and lowering CTX-I was significantly greater than that of valsartan at 8 weeks. In addition, after 4 weeks, QSYQ enhanced the protein and mRNA expression of MMP-1 and TIMP-1, and its effect on highering TIMP-1 was significantly greater than that of valsartan, whereas there was no significant difference in the expression of myocardial MMP-1 or TIMP-1 at 8 weeks. QSYQ reduced the ratio of MMP-1/TIMP-1, which further reduced over time, and the effect of QYSQ was significantly greater than that of valsartan after 4 weeks. This study provides evidence that QSYQ can reduce the rate of myocardial collagen synthesis and degradation. It also effectively improved the degree of myocardial fibrosis in experimental autoimmune myocarditis rats and it had a tendency to have a greater effect with longer treatment duration, which is related to the mechanism of regulation of MMP-1 and TIMP-1 expression in the myocardial rat.
Sigma-Aldrich
Freund′s Adjuvant, Complete, cell suspension
Sigma-Aldrich
Myosin, Calcium activated from porcine heart, buffered aqueous glycerol solution, 0.1-0.5 units/mg protein (biuret)