Merck
  • DOPAL derived alpha-synuclein oligomers impair synaptic vesicles physiological function.
DOPAL derived alpha-synuclein oligomers impair synaptic vesicles physiological function.
Scientific reports (2017-01-14)
N Plotegher, G Berti, E Ferrari, I Tessari, M Zanetti, L Lunelli, E Greggio, M Bisaglia, M Veronesi, S Girotto, M Dalla Serra, C Perego, L Casella, L Bubacco
ABSTRACT
Parkinson's disease is a neurodegenerative disorder characterized by the death of dopaminergic neurons and by accumulation of alpha-synuclein (aS) aggregates in the surviving neurons. The dopamine catabolite 3,4-dihydroxyphenylacetaldehyde (DOPAL) is a highly reactive and toxic molecule that leads to aS oligomerization by covalent modifications to lysine residues. Here we show that DOPAL-induced aS oligomer formation in neurons is associated with damage of synaptic vesicles, and with alterations in the synaptic vesicles pools. To investigate the molecular mechanism that leads to synaptic impairment, we first aimed to characterize the biochemical and biophysical properties of the aS-DOPAL oligomers; heterogeneous ensembles of macromolecules able to permeabilise cholesterol-containing lipid membranes. aS-DOPAL oligomers can induce dopamine leak in an in vitro model of synaptic vesicles and in cellular models. The dopamine released, after conversion to DOPAL in the cytoplasm, could trigger a noxious cycle that further fuels the formation of aS-DOPAL oligomers, inducing neurodegeneration.
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
m-Aminophenylboronic acid–Agarose, saline suspension