Merck
The Aurora kinases: role in cell transformation and tumorigenesis.
Cancer metastasis reviews (2003-07-30)
Hiroshi Katayama, William R Brinkley, Subrata Sen
ABSTRACT
Aurora kinases representing a novel family of serine/threonine kinases have been identified as key regulators of the mitotic cell division process. The three members of this kinase family, identified so far, referred to as Aurora-A, Aurora-B and Aurora-C kinases, are close homologues of the prototypic yeast Ipll and Drosophila aurora kinases, which are known to be involved in the regulation of centrosome function, bipolar spindle assembly and chromosome segregation processes. All three members of the mammalian kinase family have a catalytic domain that is highly conserved with a short C-terminal domain and an N-terminal domain of varying sizes. Following their discovery about five years ago, extensive research has focused on understanding the biological roles of these kinases and elucidation of their pathways, which regulate cell proliferation and maintenance of normal cellular phenotypes. Significant interest in the subject was generated since all three Aurora kinases family members were reported to be overexpressed in many human cancers, and elevated expression has been correlated with chromosomal instability and clinically aggressive disease in some instances. Ectopic overexpression of one member of the family, Aurora-A, was shown to induce oncogenic transformation in cells. Unlike most other putative oncogenes identified, so far, members of this kinase family are expressed and active at the highest level during G2-M phase of the cell cycle. Aurora kinases are localized at the centrosomes of interphase cells, at the poles of the bipolar spindle and in the midbody of the mitotic apparatus. Substrates identified for the Aurora-A and Aurora-B kinases, include a kinesin-like motor protein, spindle apparatus proteins, histone H3 protein, kinetochore protein and the tumor suppressor protein p53. Identification of Aurora kinases as RasGAP Src homology 3 domain binding protein, also implicates these kinases as potential effectors in the Ras pathway relevant to oncogenesis. Abnormal elevated expression of Aurora kinases detected in human cancer cells could help explain the underlying biological mechanisms responsible for the development of many cellular phenotypes associated with malignant cells. Identification of these mechanisms offers the possibility of designing novel targeted therapies for cancer in the future.
Sigma-Aldrich
AURORA A, active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution