MilliporeSigma
  • A bidirectional promoter connects the p14.5 gene to the gene for RNase P and RNase MRP protein subunit hPOP1.
A bidirectional promoter connects the p14.5 gene to the gene for RNase P and RNase MRP protein subunit hPOP1.
Biochemical and biophysical research communications (1998-02-12)
G Schmiedeknecht, C Büchler, G Schmitz
ABSTRACT
We have identified the functional promoter of the translational inhibitor p14.5, the human homologue to a rat perchloric acid-soluble protein (PSP), a mouse heat-responsive protein (Hrp12) and a goat tumor antigen (UK114). Sequence analysis revealed a GC-rich promoter with several consensus sequences for transcription factors, but no TATA- and CAAT-box. To confirm promoter activity, DNA fragments of the p14.5 5'-flanking region were ligated in front of the luciferase gene and were transfected into HeLa and HepG2 cells. A minimal promoter between nt -104 and nt +88 relative to the transcription start site was responsible for basal activity. Furthermore, we observed a head-to-head orientation of p14.5 to the gene for the protein subunit of RNase P and MRP ribonucleoproteins (hPOP1). Luciferase assays with fragments of the hPOP1 5'-flanking region revealed a minimal promoter between nt -20 and nt +98 relative to the start of transcription. These data indicate that the 102 bp region between p14.5 and hPOP1 can act as a bidirectional promoter. The p14.5-hPOP1-cluster was mapped to chromosome 8q22 using in situ hybridization technique.