Merck
  • The anti-apoptotic Bcl-2 family protein A1/Bfl-1 regulates neutrophil survival and homeostasis and is controlled via PI3K and JAK/STAT signaling.
The anti-apoptotic Bcl-2 family protein A1/Bfl-1 regulates neutrophil survival and homeostasis and is controlled via PI3K and JAK/STAT signaling.
Cell death & disease (2016-02-20)
J Vier, M Groth, M Sochalska, S Kirschnek
ABSTRACT
Neutrophil granulocytes are innate effector cells of the first line of defense against pyogenic bacteria. Neutrophil lifespan is short, is prolonged by pro-inflammatory stimuli, controls functionality of the cells and can determine tissue damage. Experimental analysis of primary neutrophils is difficult because of their short lifespan and lack of possibilities of genetic manipulation. The Hoxb8 system of neutrophil differentiation from immortalized progenitor cells offers the advantage of unlimited production of neutrophils in vitro as well as easy genetic modification. We here use this system to analyze the role of the poorly characterized anti-apoptotic B-cell lymphoma protein 2 (Bcl-2) family member A1/Bfl-1 (Bcl-2-related protein A1) for survival and homeostasis of neutrophils and of neutrophil progenitors. Low constitutive mRNA and protein expression of A1 was detected, while A1 was transiently upregulated early during differentiation. Pro-inflammatory stimuli caused strong, mainly transcriptional, A1 upregulation, in contrast to posttranscriptional regulation of Mcl-1 (induced myeloid leukemia cell differentiation protein). Inhibitor studies showed that phosphoinositide-3 kinase (PI3K)/Akt and Janus kinase (JAK)/signal transducer and activator of transcription (STAT) is required for A1 expression and survival of progenitors and mature neutrophils. ShRNA-mediated constitutive A1 knockdown (KD) impaired maintenance of progenitors. ShRNA experiments further showed that A1 was required early during neutrophil differentiation as well as in mature neutrophils upon pro-inflammatory stimulation. Our data further indicate differential regulation of the two anti-apoptotic proteins A1 and Mcl-1. Relevant findings were confirmed in primary human neutrophils. Our data indicate that A1, in addition to the well-established Mcl-1, substantially contributes to neutrophil survival and homeostasis. A1 may thus be a promising target for anti-inflammatory therapy.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O55:B5, purified by gel-filtration chromatography