Merck
  • Synthesis and in vitro and in vivo characteristics of an iodinated analogue of the beta-adrenoceptor antagonist carazolol.
Synthesis and in vitro and in vivo characteristics of an iodinated analogue of the beta-adrenoceptor antagonist carazolol.
Journal of medicinal chemistry (1996-08-16)
E A Dubois, J C van den Bos, T Doornbos, P A van Doremalen, G A Somsen, J A Vekemans, A G Janssen, H D Batink, G J Boer, M Pfaffendorf, E A van Royen, P A van Zwieten
ABSTRACT
A new (radio)iodinated, beta-adrenoceptor ligand, (S)-(-)-4-[3-[(1,1-dimethyl-3-iodo-(2E)-propenyl)-amino]-2- hydroxypropoxy]carbazole (CYBL8E, 1), was prepared. 1 is an iodinated analogue of the high-affinity beta-adrenoceptor antagonist carazolol (2). The asymmetric synthesis was achieved in four steps starting from 4-hydroxycarbazole. The iodine-123-labeled form was obtained by an iododestannylation reaction with [123I]NaI in the presence of H2O2. Using classical in vitro displacement experiments with membrane fractions of cardiac left ventricular muscle, 1 proved to have a high affinity for the receptor (Ki = 0.31 +/- 0.03). Biodistribution studies performed in New Zealand white rabbits demonstrated the specificity of the binding in vivo to the receptor. Uptake of [123I]1 was reduced significantly in both atrial muscle, left ventricular muscle, frontal cortex, cerebellum, and striatum, by the pretreatment of the animals with different beta-adrenoceptor antagonists. In conclusion, 1 is a potent nonselective beta-adrenoceptor antagonist, which binds specifically to the beta-adrenoceptor in vivo, and is therefore a promising radioligand for the imaging of beta-adrenoceptors using single photon emission computerized tomography.