Merck
Synthesis of ZnO nanoparticles and their ink-jetting behavior.
Journal of nanoscience and nanotechnology (2012-07-05)
Mohammad Vaseem, Kil-Mok Lee, Jin-Koog Shin, Yoon-Bong Hahn
ABSTRACT
Zinc oxide (ZnO) nanoparticles were synthesized by ultrasonically-assisted solution process without using any surfactants. The as-synthesized spherical ZnO nanoparticles with diameter of 20 +/- 5 nm possessed crystalline nature with wurtzite hexagonal phase and showed a blue-shifted near band-edge ultra-violet absorption at -340 nm due to quantum confinement effect of ZnO nanoparticles. The as-synthesized ZnO nanoparticles were further formulated as an ink for ink-jet printing application. Jetting and writing of various line patterns on Si and ITO substrates were demonstrated using the as-formulated ZnO ink. UV-treated substrates showed an improvement in the fixation of ink on substrate due to decreased contact angle with controlled surface wettability.
Sigma-Aldrich
Zinc oxide nanoparticle ink, 2.5 wt. %, viscosity 10 cP, work function -4.3eV