Merck
  • Molecular characterization of porcine SARM1 and its role in regulating TLRs signaling during highly pathogenic porcine reproductive and respiratory syndrome virus infection in vivo.
Molecular characterization of porcine SARM1 and its role in regulating TLRs signaling during highly pathogenic porcine reproductive and respiratory syndrome virus infection in vivo.
Developmental and comparative immunology (2012-03-01)
Xiang Zhou, Tengfei Jiang, Xiaochuan Du, Ping Zhou, Zhihua Jiang, Jennifer J Michal, Bang Liu
ABSTRACT
Toll-like receptors (TLRs) are important pattern-recognition receptors (PRRs) that trigger innate immune response and mediate acquired immunity. Evidence has shown that SARM1 (sterile-α and TIR motif containing protein 1) is one of five TIR domain-containing adaptor proteins involved in TLRs signaling transduction. In the present study, a full-length cDNA sequence was cloned for the porcine SARM1 gene, which contains nine exons. Using the radiation hybrid mapping approach, we assigned the porcine gene to SSC12 q13. Under the normal condition, porcine SARM1 was highly expressed in brain and spleen. Polyinosinic-polycytidylic acid (poly (I:C)) weakly induced the porcine SARM1 expression in the early stimulation. We found that porcine SARM1 protein is localized in mitochondria and attenuates NF-κB activation induced by stimulation and infection. The quantitative real-time PCR (Q-PCR) analysis showed that the expression of porcine SARM1 significantly decreased in several tissues of Tongcheng pigs infected with highly pathogenic porcine reproductive and respiratory syndrome virus (HP-PRRSV). Gene-interaction network analysis for porcine SARM1 in porcine alveolar macrophages (PAMs) showed that down-regulation of SARM1 gene in infected Tongcheng pig may modulate TRIF-depend TLRs signaling and regulate the expression of disease-resistant genes and inflammatory genes. Our findings provide evidence that porcine SARM1 may play an important role in immune regulation with PRRSV infection.