Merck
  • Flavopiridol, a cyclin-dependent kinase inhibitor, enhances radiosensitivity of ovarian carcinoma cells.
Flavopiridol, a cyclin-dependent kinase inhibitor, enhances radiosensitivity of ovarian carcinoma cells.
Cancer research (2003-06-18)
Uma Raju, Eiko Nakata, Kathy A Mason, K Kian Ang, Luka Milas
ABSTRACT
Flavopiridol, a cyclin-dependent kinase (cdk) inhibitor, can cause cell cycle arrest, induce apoptosis in cancer cells, and inhibit tumor cell growth in vivo. The present study investigated the in vitro radiosensitizing effect of flavopiridol and the underlying molecular mechanisms in a murine ovarian cancer cell line, OCA-I. Flavopiridol inhibited cell growth in a dose-dependent manner and enhanced cell radiosensitivity assessed by the clonogenic cell survival assay. A flavopiridol dose of 300 nM, given for 1 day, enhanced radiosensitivity by a factor of 2.1. Clonogenic cell survival after split-dose radiation showed that flavopiridol inhibited repair from radiation damage. In addition, flavopiridol treatment (300 nM, 1 day) resulted in decreased levels of Ku70 and Ku86 proteins that play a role in DNA repair processes, suggesting that DNA repair processes may have been disrupted by this agent. Flow cytometry analysis showed that flavopiridol (300 nM, 1 day) accumulated the cells in G(1) and G(2) phases, with a significant reduction in the S phase component. This cell cycle redistribution is likely another mechanism underlying flavopiridol-induced cell radiosensitivity. Flavopiridol down-regulated cyclin D1 and cyclin E protein levels and also inhibited phosphorylation of retinoblastoma protein, which is inconsistent with the observed cell cycle arrest. Among the cdks tested, cdk-9, the catalytic subunit of positive transcription elongation factor b, was significantly down-regulated by flavopiridol, suggesting that flavopiridol may modulate cellular transcription processes. Furthermore, flavopiridol on its own induced apoptosis in the OCA-I cells, whereas in combination with radiation, exerted no additional increase in apoptosis. Taken together, our data show that flavopiridol strongly augmented the response of ovarian carcinoma cells to radiation and that the underlying mechanisms included inhibition of sublethal DNA damage repair and cell cycle redistribution. At the molecular level, transcriptional regulation by flavopiridol may have been involved.
Sigma-Aldrich
Flavopiridol hydrochloride hydrate, ≥98% (HPLC), powder