Merck
  • RNA interference-mediated FANCF silencing sensitizes OVCAR3 ovarian cancer cells to adriamycin through increased adriamycin-induced apoptosis dependent on JNK activation.
RNA interference-mediated FANCF silencing sensitizes OVCAR3 ovarian cancer cells to adriamycin through increased adriamycin-induced apoptosis dependent on JNK activation.
Oncology reports (2013-02-27)
Miao He, Hai-Gang Sun, Jun-Ying Hao, Yan-Lin Li, Jian-Kun Yu, Yuan-Yuan Yan, Lin Zhao, Na Li, Yan Wang, Xue-Feng Bai, Zhao-Jin Yu, Zhi-Hong Zheng, Xiao-Yi Mi, En-Hua Wang, Min-Jie Wei
ABSTRACT
In the present study, we downregulated FANCF expression by small interfering RNA (siRNA) in OVCAR ovarian cancer cells to address the effects of decreased FANCF expression on the function of the Fanconi anemia (FA)/breast cancer susceptibility gene (BRCA) pathway. Furthermore, we investigated whether this method increases the sensitivity of OVCAR3 cells to adriamycin (ADM) and the possible mechanism(s). We found that silencing of FANCF inactivated the FA/BRCA pathway by decreasing the monoubiquitination and focus formation of FANCD2 and reduced the function of the FA/BRCA pathway, resulting in the inhibition of cell proliferation, increased cell apoptosis and DNA damage in OVCAR3 cells. Moreover, we observed that silencing of FANCF enhanced the antiproliferative effect of ADM in OVCAR3 cells and increased ADM intracellular accumulation consequently sensitizing OVCAR3 cells to ADM. Furthermore, silencing of FANCF increased cell apoptosis of OVCAR3 cells which was caused by decreased mitochondrial membrane potential (MMP)-induced DNA damage, activated Jun N-terminal kinase (JNK), increased release of cytochrome c, increased expression of cleaved caspase-3 and poly(ADP-ribose) polymerase (PARP) dependent on JNK activation following treatment of ADM. Collectively, we confirm that silencing of FANCF sensitizes OVCAR3 ovarian cancer cells to ADM, suggesting that FANCF may serve as a potential target for therapeutic strategies in the treatment of ovarian cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting human FANCF