Merck
  • Clinical significance of E2F1 protein expression in non-small cell lung cancer.
Clinical significance of E2F1 protein expression in non-small cell lung cancer.
Experimental hematology & oncology (2012-12-06)
Jung-Jyh Hung, Chung-Tsen Hsueh, Kuan-Hua Chen, Wen-Hu Hsu, Yu-Chung Wu
ABSTRACT
The transcription factor E2F1 has been implicated in cell cycle control and DNA damage response. Paradoxically, E2F1 can promote apoptosis and function as tumor suppressor. In non-small cell lung cancer (NSCLC), there are conflicting data for clinical significance of E2F1 expression. In this study, we investigated the protein expression of E2F1 in patients with stage I-III NSCLC, and its correlation with clinical outcome. 56 paired adjacent non-tumor/tumor matched samples were prospectively obtained from patients undergoing surgery for stage I-III NSCLC at Taipei Veterans General Hospital. The protein expression of E2F1 was determined by Western blot analysis. The levels of E2F1 protein were significantly higher in tumor samples than in non-tumor lung specimens (P = 0.008). Overexpression of E2F1 was defined as a more than 2-fold expression in the tumorous sample compared with the corresponding nontumorous one, and was noted in 21 patients (37.5%). There was no significant difference in overall survival (P = 0.44) or probability of freedom from recurrence (P = 0.378) between patients with E2F1 overexpression vs. non-overexpressors. Additionally, there was no significant association between E2F1 overexpression and any clinicopathologic parameter such as histological type, stage, or angiolymphatic invasion of tumor. E2F1 protein is frequently overexpressed in NSCLC. There is no correlation between E2F1 protein expression and clinical outcome such as survival and freedom from progression.
Sigma-Aldrich
MISSION® esiRNA, targeting human E2F1