Merck
  • Tool compounds robustly increase turnover of an artificial substrate by glucocerebrosidase in human brain lysates.
Tool compounds robustly increase turnover of an artificial substrate by glucocerebrosidase in human brain lysates.
PloS one (2015-03-13)
Zdenek Berger, Sarah Perkins, Claude Ambroise, Christine Oborski, Matthew Calabrese, Stephen Noell, David Riddell, Warren D Hirst
ABSTRACT
Mutations in glucocerebrosidase (GBA1) cause Gaucher disease and also represent a common risk factor for Parkinson's disease and Dementia with Lewy bodies. Recently, new tool molecules were described which can increase turnover of an artificial substrate 4MUG when incubated with mutant N370S GBA1 from human spleen. Here we show that these compounds exert a similar effect on the wild-type enzyme in a cell-free system. In addition, these tool compounds robustly increase turnover of 4MUG by GBA1 derived from human cortex, despite substantially lower glycosylation of GBA1 in human brain, suggesting that the degree of glycosylation is not important for compound binding. Surprisingly, these tool compounds failed to robustly alter GBA1 turnover of 4MUG in the mouse brain homogenate. Our data raise the possibility that in vivo models with humanized glucocerebrosidase may be needed for efficacy assessments of such small molecules.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Avanti
C18 Glucosyl(β) Ceramide (d18:1/18:0), Avanti Research - A Croda Brand
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Anti-Glucocerebrosidase (C-terminal) antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Taurocholic acid sodium salt hydrate, ≥95% (HPLC)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 1, DMSO solution
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution