Merck
Total synthesis and structural revision of (+)-amphidinolide W.
Journal of the American Chemical Society (2004-03-25)
Arun K Ghosh, Gangli Gong
ABSTRACT
An enantioselective first total syntheis of amphidinolide W (2) and a revision of its C6 absolute stereochemistry (1) are described. Amphidinolide W (1), a 12-membered macrolide isolated from Amphidinium sp., has shown potent antitumor properties against a variety of NCI tumor cell lines. The synthesis is convergent, and four of the five chiral centers were derived through asymmetric synthesis. The synthesis features Sharpless asymmetric dihydroxylation, diastereoselective alkylation, efficient cross metathesis of functionalized substrates, and novel functional group transformations using selective lipase-catalyzed hydrolysis of the primary acetate group. Of particular note, the C6 absolute stereochemistry of amphidinolide W (1) has now been revised through our current synthesis.
Sigma-Aldrich
(S)-4-Benzyl-2-oxazolidinone, 99%