Merck
Inflammatory signaling compromises cell responses to interferon alpha.
Oncogene (2011-06-15)
W-C Huangfu, J Qian, C Liu, J Liu, A E Lokshin, D P Baker, H Rui, S Y Fuchs
ABSTRACT
Interferon alpha (IFNα) is widely used for treatment of melanoma and certain other malignancies. This cytokine as well as the related IFNβ exerts potent anti-tumorigenic effects; however, their efficacy in patients is often suboptimal. Here, we report that inflammatory signaling impedes the effects of IFNα/β. Melanoma cells can secrete pro-inflammatory cytokines that inhibit cellular responses to IFNα/β via activating the ligand-independent pathway for the phosphorylation and subsequent ubiquitination and accelerated degradation of the IFNAR1 chain of type I IFN receptor. Catalytic activity of the p38 protein kinase was required for IFNAR1 downregulation and inhibition of IFNα/β signaling induced by proinflammatory cytokines such as interleukin 1 (IL-1). Activation of p38 kinase inversely correlated with protein levels of IFNAR1 in clinical melanoma specimens. Inhibition of p38 kinase augmented the inhibitory effects of IFNα/β on cell viability and growth in vitro and in vivo. The roles of inflammation and p38 protein kinase in regulating cellular responses to IFNα/β in normal and tumor cells are discussed.
Sigma-Aldrich
CGP 57380, ≥98% (HPLC)