- Notch signal transduction: a real rip and more.
Notch signal transduction: a real rip and more.
Current opinion in genetics & development (2000-07-13)
G Weinmaster
PMID10889061
ABSTRACT
The Notch signaling pathway functions in a wide variety of processes that regulate tissue patterning and morphogenesis in developing vertebrates and invertebrates. Research on the mechanism of ligand-induced Notch signal transduction has revealed a novel and essential element in the signal cascade. Some recent findings support a model in which sequential proteolytic cleavage serves to regulate Notch signal transduction.