Merck
  • A high-performance liquid chromatography method for measurement of oxidized glutathione in biological samples.
A high-performance liquid chromatography method for measurement of oxidized glutathione in biological samples.
Analytical biochemistry (1994-03-01)
M Asensi, J Sastre, F V Pallardó, J García de la Asunción, J M Estrela, J Viña
ABSTRACT
A high-performance liquid chromatography method to determine oxidized glutathione (GSSG) in biological samples with ultraviolet-visible detection using N-ethylmaleimide to prevent reduced glutathione (GSH) oxidation is described. Previous methods based on high-performance liquid chromatography to quantitative GSH and GSSG are unsuitable for determining GSSG in biological samples. This is due to GSH oxidation during sample processing. N-Ethylmaleimide, but not iodacetic acid, prevents this oxidation. Blood GSH oxidation measured by the widely used method of Reed et al. (Anal. Biochem. 106, 55-62, 1980) can be as high as 24 +/- 6% (n = 6). When blood samples were assayed by our procedure, GSH oxidation was only 0.13 +/- 0.28% (n = 5). GSH can be determined enzymatically, i.e., with glutathione-S-transferase, but perchloric acid should not be used to deproteinize samples. Trichloroacetic acid (15% final concentration) may be used. This method allows an accurate calculation of the GSH/GSSG ratio, which is important for determining oxidative stress in tissues in various pathophysiological situations.