Merck
  • Renin inhibitors. Synthesis of transition-state analogue inhibitors containing phosphorus acid derivatives at the scissile bond.
Renin inhibitors. Synthesis of transition-state analogue inhibitors containing phosphorus acid derivatives at the scissile bond.
Journal of medicinal chemistry (1989-07-01)
M C Allen, W Fuhrer, B Tuck, R Wade, J M Wood
ABSTRACT
The synthesis of five amino phosphorus derivatives, 1a-e, is described. The derivatives were incorporated into a series (18) of analogues of the 5-14 portion of angiotensinogen, in most cases at the scissile Leu-Val bond. The resultant compounds were tested in vitro for their ability to inhibit human plasma renin. Replacement of the scissile bond with the phosphinic analogue of Leu10-Val11 (1b) gave the most potent inhibitors, having IC50 = 7.5 x 10(-8) M for H-Pro-His-Pro-Phe-His-(1b)-Ile-His-Lys-OH and IC50 = 1.0 x 10(-7) M for Z-Arg-Arg-Pro-Phe-His-(1b)-Ile-His-NH2. The shorter phosphonic acid sequence Z-Pro-Phe-His-(1d) retained biological activity with an IC50 = 6.4 x 10(-6) M.
Sigma-Aldrich
(1R)-(+)-(1-Amino-2-methylpropyl)phosphonic acid, 98%