Merck
  • Lysophosphatidic acid activates Arf6 to promote the mesenchymal malignancy of renal cancer.
Lysophosphatidic acid activates Arf6 to promote the mesenchymal malignancy of renal cancer.
Nature communications (2016-02-09)
Shigeru Hashimoto, Shuji Mikami, Hirokazu Sugino, Ayumu Yoshikawa, Ari Hashimoto, Yasuhito Onodera, Shotaro Furukawa, Haruka Handa, Tsukasa Oikawa, Yasunori Okada, Mototsugu Oya, Hisataka Sabe
ABSTRACT
Acquisition of mesenchymal properties by cancer cells is critical for their malignant behaviour, but regulators of the mesenchymal molecular machinery and how it is activated remain elusive. Here we show that clear cell renal cell carcinomas (ccRCCs) frequently utilize the Arf6-based mesenchymal pathway to promote invasion and metastasis, similar to breast cancers. In breast cancer cells, ligand-activated receptor tyrosine kinases employ GEP100 to activate Arf6, which then recruits AMAP1; and AMAP1 then binds to the mesenchymal-specific protein EPB41L5, which promotes epithelial-mesenchymal transition and focal adhesion dynamics. In renal cancer cells, lysophosphatidic acid (LPA) activates Arf6 via its G-protein-coupled receptors, in which GTP-Gα12 binds to EFA6. The Arf6-based pathway may also contribute to drug resistance. Our results identify a specific mesenchymal molecular machinery of primary ccRCCs, which is triggered by a product of autotaxin and it is associated with poor outcome of patients.
Sigma-Aldrich
Anti-LPAR2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Pepstatin A, microbial, ≥90% (HPLC)
Sigma-Aldrich
Leupeptin hydrochloride, microbial, ≥90% (HPLC)
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized powder, 3-7 TIU/mg solid
Sigma-Aldrich
E-64, protease inhibitor
Sigma-Aldrich
Anti-PSD4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution