Merck
  • A rapid and effective synthesis of propylene carbonate using a supercritical CO2-ionic liquid system.
A rapid and effective synthesis of propylene carbonate using a supercritical CO2-ionic liquid system.
Chemical communications (Cambridge, England) (2003-05-13)
Hajime Kawanami, Akiyoshi Sasaki, Keitaro Matsui, Yutaka Ikushima
ABSTRACT
The synthesis of propylene carbonate from propylene oxide and carbon dioxide under supercritical conditions in the presence of 1-octyl-3-methylimidazolium tetrafluoroborate was achieved in nearly 100% yield and 100% selectivity within 5 minutes, whose TOF value is 77 times larger than those so far reported.
Sigma-Aldrich
Propylene carbonate, anhydrous, 99.7%