Merck
  • A novel self-assembling synthesis and crystal structure of 40-membered macrocyclic complex containing eight-tin.
A novel self-assembling synthesis and crystal structure of 40-membered macrocyclic complex containing eight-tin.
Dalton transactions (Cambridge, England : 2003) (2004-09-24)
Chunlin Ma, Jiafeng Sun
ABSTRACT
The self-assembly reaction of 2-mercapto-4-methyl-5-thiazoleacetic acid, sodium methoxide and dimethyltin dichloride in moist dichloromethane affords a novel eight-tin macrocyclic complex, which is a 40-membered macrocycle containing two distannoxane ladders.
Sigma-Aldrich
2-Mercapto-4-methyl-5-thiazoleacetic acid, 98%