Merck
  • DNA damage-inducible gene p33ING2 negatively regulates cell proliferation through acetylation of p53.
DNA damage-inducible gene p33ING2 negatively regulates cell proliferation through acetylation of p53.
Proceedings of the National Academy of Sciences of the United States of America (2001-08-02)
M Nagashima, M Shiseki, K Miura, K Hagiwara, S P Linke, R Pedeux, X W Wang, J Yokota, K Riabowol, C C Harris
ABSTRACT
The p33ING1 protein is a regulator of cell cycle, senescence, and apoptosis. Three alternatively spliced transcripts of p33ING1 encode p47ING1a, p33ING1b, and p24ING1c. We cloned an additional ING family member, p33ING2/ING1L. Unlike p33ING1b, p33ING2 is induced by the DNA-damaging agents etoposide and neocarzinostatin. p33ING1b and p33ING2 negatively regulate cell growth and survival in a p53-dependent manner through induction of G(1)-phase cell-cycle arrest and apoptosis. p33ING2 strongly enhances the transcriptional-transactivation activity of p53. Furthermore, p33ING2 expression increases the acetylation of p53 at Lys-382. Taken together, p33ING2 is a DNA damage-inducible gene that negatively regulates cell proliferation through activation of p53 by enhancing its acetylation.