Merck
  • Inhalative IL-10 treatment after bilateral femoral fractures affect pulmonary inflammation in mice.
Inhalative IL-10 treatment after bilateral femoral fractures affect pulmonary inflammation in mice.
Annals of anatomy = Anatomischer Anzeiger : official organ of the Anatomische Gesellschaft (2015-03-25)
Philipp Lichte, Roman Pfeifer, Philipp Kobbe, Mersedeh Tohidnezhad, Thomas Pufe, Khalid Almahmoud, Frank Hildebrand, Hans-Christoph Pape
ABSTRACT
Musculoskeletal injuries induce systemic inflammation which often impairs lung function contributing to morbidity. IL-10 has been shown to have a beneficial effect on immune dysfunction and organ damage after different traumatic insults. We sought to investigate the effect of inhalative IL-10 administration on the systemic and pulmonary inflammatory response in a small animal model of bilateral femoral fracture. Male C57/BL6 mice (6 animals per group) were subjected to bilateral femoral fracture and intramedullary nailing followed by inhalative administration of either 50μL PBS (Fx group) or 50μg/kg recombinant mouse IL-10 dissolved in 50μL PBS (FxIL-10 group). All animals were sacrificed at 6, 24, or 72h after fracture induction. Blood samples were collected and analyzed for IL-6, IL-10, KC, and MCP-1 (CCL2) plasma concentrations by Bio-Plex Pro™ assays. Pulmonary infiltration by neutrophils was assessed by myeloperoxidase (MPO) activity (ELISA) and histological analysis of lung tissue. Pulmonary ICAM-1 expression (immunohistochemistry), and pulmonary IL-6 levels (ELISA) were determined. Inhalative IL-10 administration showed a decrease in the pulmonary infiltration by neutrophils. A significant decrease in the expression of the adhesion molecule ICAM-1 after local IL-10 application was observed. In contrast, local IL-10 administration did not show a significant effect on the systemic inflammatory response. Our findings suggest that inhalative IL-10 administration may beneficially modulate the pulmonary microenvironment, in which IL-10 effect on the local ICAM-1 expression seems to play a central role.
Sigma-Aldrich
Interleukin-10 from mouse, >97% (SDS-PAGE), recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)