Merck
  • Effects of vitamin D supplementation on intestinal permeability, cathelicidin and disease markers in Crohn's disease: Results from a randomised double-blind placebo-controlled study.
Effects of vitamin D supplementation on intestinal permeability, cathelicidin and disease markers in Crohn's disease: Results from a randomised double-blind placebo-controlled study.
United European gastroenterology journal (2015-07-03)
Tara Raftery, Adrian R Martineau, Claire L Greiller, Subrata Ghosh, Deirdre McNamara, Kathleen Bennett, Jon Meddings, Maria O'Sullivan
ABSTRACT
Vitamin D (vitD) supplementation may prolong remission in Crohn's disease (CD); however, the clinical efficacy and mechanisms are unclear. To determine changes in intestinal permeability (IP), antimicrobial peptide (AMP) concentrations and disease markers in CD, in response to vitD supplementation. In a double-blind randomised placebo-controlled study, we assigned 27 CD patients in remission to 2000 IU/day vitD or placebo for 3 mos. We determined IP, plasma cathelicidin (LL-37 in ng/mL), human-beta-defensin-2 (hBD2 in pg/mL), disease activity (Crohn's Disease Activity Index (CDAI)), C-reactive protein (CRP in mg/L), fecal calprotectin (µg/g), Quality of Life (QoL) and serum 25-hydroxyvitamin D (25(OH)D in nmol/L) at 0 and 3 mos. At 3 mos., 25(OH)D concentrations were significantly higher in those whom were treated (p < 0.001). Intra-group analysis showed increased LL-37 concentrations (p = 0.050) and maintenance of IP measures in the treated group. In contrast, in the placebo group, the small bowel (p = 0.018) and gastro-duodenal permeability (p = 0.030) increased from baseline. At 3 mos., patients with 25(OH)D ≥ 75 nmol/L had significantly lower CRP (p = 0.019), higher QoL (p = 0.037), higher LL-37 concentrations (p < 0.001) and non-significantly lower CDAI scores (p = 0.082), compared to those with levels <75 nmol/L. Short-term treatment with 2000 IU/day vitD significantly increased 25(OH)D levels in CD patients in remission and it was associated with increased LL-37 concentrations and maintenance of IP. Achieving 25(OH)D ≥ 75 nmol/l was accompanied by higher circulating LL-37, higher QoL scores and reduced CRP. Registered at ClinicalTrials.gov (NCT01792388).
Sigma-Aldrich
Sucralose, ≥98.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Lactulose, ≥95% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Lactulose, ≥98.0% (HPLC)
Pricing and availability is not currently available.