Merck
  • Cost-Effective and Highly Photoresponsive Nanophosphor-P3HT Photoconductive Nanocomposite for Near-Infrared Detection.
Cost-Effective and Highly Photoresponsive Nanophosphor-P3HT Photoconductive Nanocomposite for Near-Infrared Detection.
Scientific reports (2015-11-17)
Yi Tong, Xinyu Zhao, Mei Chee Tan, Rong Zhao
ABSTRACT
The advent of flexible optoelectronic devices has accelerated the development of semiconducting polymeric materials. We seek to replace conventional expensive semiconducting photodetector materials with our cost-effective composite system. We demonstrate in this work the successful fabrication of a photoconductive composite film of poly(3-hexylthiophene-2,5-diyl) (P3HT) mixed with NaYF4:Yb,Er nanophosphors that exhibited a ultrahigh photoresponse to infrared radiation. The high photocurrent measured was enabled by the unique upconversion properties of NaYF4:Yb,Er nanophosphors, where low photon energy infrared excitations are converted to high photon energy visible emissions that are later absorbed by P3HT. Here we report, a significant 1.10 × 10(5) times increment of photocurrent from our photoconductive composite film upon infrared light exposure, which indicates high optical-to-electrical conversion efficiency. Our reported work lays the groundwork for the future development of printable, portable flexible and functional photonic composites for light sensing and harvesting, photonic memory devices, and phototransistors.
Sigma-Aldrich
Ytterbium, powder, ≥99.9% trace rare earth metals basis
Sigma-Aldrich
Yttrium, chips, 99.9% trace rare earth metals basis
Yttrium, rod, 50mm, diameter 6.35mm, cast, 99%
Sigma-Aldrich
Yttrium sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.9% trace metals basis
Ytterbium, rod, 50mm, diameter 2.0mm, cast, 99%
Ytterbium, lump, 50g, max. size 25mm, 99.9%
Ytterbium, rod, 50mm, diameter 10.0mm, cast, 99%
Ytterbium, rod, 50mm, diameter 6.35mm, cast, 99%
Ytterbium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Yttrium, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99%
Ytterbium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Yttrium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Yttrium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Ytterbium, lump, 100g, max. size 25mm, 99.9%
Yttrium, foil, 25mm disks, thickness 0.50mm, as rolled, 99%
Ytterbium, lump, 25g, max. size 25mm, 99.9%
Yttrium, rod, 100mm, diameter 12.5mm, cast, 99%
Ytterbium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Ytterbium, rod, 100mm, diameter 6.35mm, cast, 99%
Yttrium, wire reel, 0.1m, diameter 0.5mm, 99.9%
Yttrium, rod, 100mm, diameter 6.35mm, cast, 99%
Yttrium, rod, 50mm, diameter 2.0mm, cast, 99%
Yttrium, wire reel, 0.05m, diameter 1.0mm, 99.9%
Yttrium, wire reel, 0.2m, diameter 1.0mm, 99.9%
Yttrium, wire reel, 0.05m, diameter 0.5mm, 99.9%
Yttrium, wire reel, 0.1m, diameter 1.0mm, 99.9%
Yttrium, rod, 50mm, diameter 12.5mm, cast, 99%
Sigma-Aldrich
1-Octadecene, ≥95.0% (GC)
Sigma-Aldrich
Oleylamine, technical grade, 70%
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)