Merck
  • Validated UPLC-MS/MS method for simultaneous determination of simvastatin, simvastatin hydroxy acid and berberine in rat plasma: Application to the drug-drug pharmacokinetic interaction study of simvastatin combined with berberine after oral administration in rats.
Validated UPLC-MS/MS method for simultaneous determination of simvastatin, simvastatin hydroxy acid and berberine in rat plasma: Application to the drug-drug pharmacokinetic interaction study of simvastatin combined with berberine after oral administration in rats.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-11-01)
Mei Liu, Xianying Su, Guofei Li, Guilian Zhao, Limei Zhao
ABSTRACT
A rapid and sensitive liquid chromatography-tandem mass spectrometric (LC-MS/MS) assay method was developed and validated for simultaneous quantification of simvastatin (SV), its metabolite simvastatin hydroxy acid (SVA) and berberine (BBR) in rat plasma. Separation was performed on Poroshell 120 EC-C18 column (4.6×50mm, 2.7μm) using gradient elution by mobile phase containing acetonitrile and 10mM ammonium acetate (pH 4.5). Polarity switch (positive-negative-positive ionization mode) was performed in a total run time of 4.0min. The lower limits of quantification (LLOQ) for SV, SVA and BBR were 0.10, 0.20 and 0.10ng/mL, respectively. The response function was established for concentration range of 0.10-100ng/mL for SV and BBR and 0.20-3000ng/mL for SVA, with a coefficient of correlation of >0.99 for all the compounds. The proposed method was applied to the drug-drug pharmacokinetic interaction study of SV combined with BBR after oral administration in rats.
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, Absolute - Acetone free
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, NMR reference standard
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Ultrapure Acetonitrile
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C