Merck
  • Intracellular sirolimus concentration is reduced by tacrolimus in human pancreatic islets in vitro.
Intracellular sirolimus concentration is reduced by tacrolimus in human pancreatic islets in vitro.
Transplant international : official journal of the European Society for Organ Transplantation (2015-06-06)
Kristine Kloster-Jensen, Nils Tore Vethe, Sara Bremer, Shadab Abadpour, Olle Korsgren, Aksel Foss, Stein Bergan, Hanne Scholz
ABSTRACT
Islet transplantation has become a promising treatment for type 1 diabetes. However, immunosuppressive drugs used today cause islet deterioration and modification strategies are necessary. But little is known about pharmacokinetics interactions and intracellular concentrations of immunosuppressive drugs in human islets. We determined the pharmacokinetics of tacrolimus and sirolimus in islets by measuring intracellular concentration after exposure alone or in combination at two different doses up to 48 h. A quantification technique established in our laboratory using a Micromass Quattro micro API MS/MS-instrument with electrospray ionization was used. Islets function was measured by oxygen consumption rates. Presence of drug transporters OATP1B1 and ABCB1 and metabolizing enzyme CYP3A4 in islets were quantified using real-time quantitative PCR. Islets incubated with tacrolimus and sirolimus had a significant decrease in intracellular concentration of sirolimus compared to sirolimus alone. Reduced intracellular sirolimus concentration was followed by increased p70S6k phosphorylation suggesting preservation of the mTOR-signaling pathway. Drug transporters OATP1B1 and ABCB1 and enzyme CYP3A4 were expressed in human islets, but were not involved in the reduced sirolimus concentration by tacrolimus. These findings provide new knowledge of the drug interaction between tacrolimus and sirolimus, suggesting that tacrolimus has an inhibitory effect on the intracellular concentration of sirolimus in human islets.
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Phenol Red, ACS reagent
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Rapamycin from Streptomyces hygroscopicus, ≥95% (HPLC), powder
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
FK-506 monohydrate, ≥98% (HPLC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)