Merck
  • Preferentially Cytotoxic Constituents of Andrographis paniculata and their Preferential Cytotoxicity against Human Pancreatic Cancer Cell Lines.
Preferentially Cytotoxic Constituents of Andrographis paniculata and their Preferential Cytotoxicity against Human Pancreatic Cancer Cell Lines.
Natural product communications (2015-09-29)
Sullim Lee, Hiroyuki Morita, Yasuhiro Tezuka
ABSTRACT
In the course of our search for anticancer agents based on a novel anti-austerity strategy, we found that the 70% EtOH extract of the crude drug Andrographis Herba (aerial parts of Andrographis paniculata), used in Japanese Kampo medicines, killed PANC-1 human pancreatic cancer cells preferentially in nutrient-deprived medium (NDM). Phytochemical investigation of the 70% EtOH extract led to the isolation of 21 known compounds consisting of six labdane-type diterpenes (11, 15, 17-19, 21), six flavones (5, 7, 10, 12, 14, 20), three flavanones (2, 6, 16), two sterols (3, 8), a fatty acid (1), a phthalate (4), a triterpene (9), and a monoterpene (13). Among them, 14-deoxy-11,12-didehydroandrographolide (17) displayed the most potent preferential cytotoxicity against PANC-1 and PSN-1 cells with PC50 values of 10.0 μM and 9.27 μM, respectively. Microscopical observation, double staining with ethidium bromide (EB) and acridine orange (AO), and flow cytometry with propidium iodide/annexin V double staining indicated that 14-deoxy-11,12-didehydroandrographolide (17) triggered apoptosis-like cell death in NDM with an amino acids and/or serum-sensitive mode.
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
1-Dodecanethiol, ≥98%
Sigma-Aldrich
Phenol Red, ACS reagent