Merck
  • Fast and sensitive detection of indels induced by precise gene targeting.
Fast and sensitive detection of indels induced by precise gene targeting.
Nucleic acids research (2015-03-11)
Zhang Yang, Catharina Steentoft, Camilla Hauge, Lars Hansen, Allan Lind Thomsen, Francesco Niola, Malene B Vester-Christensen, Morten Frödin, Henrik Clausen, Hans H Wandall, Eric P Bennett
ABSTRACT
The nuclease-based gene editing tools are rapidly transforming capabilities for altering the genome of cells and organisms with great precision and in high throughput studies. A major limitation in application of precise gene editing lies in lack of sensitive and fast methods to detect and characterize the induced DNA changes. Precise gene editing induces double-stranded DNA breaks that are repaired by error-prone non-homologous end joining leading to introduction of insertions and deletions (indels) at the target site. These indels are often small and difficult and laborious to detect by traditional methods. Here we present a method for fast, sensitive and simple indel detection that accurately defines indel sizes down to ±1 bp. The method coined IDAA for Indel Detection by Amplicon Analysis is based on tri-primer amplicon labelling and DNA capillary electrophoresis detection, and IDAA is amenable for high throughput analysis.
Sigma-Aldrich
Ethyl methyl carbonate, 99%
Sigma-Aldrich
Ethyl methyl carbonate, 98%
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)