Merck
  • Long-term Benefit of PD-L1 Blockade in Lung Cancer Associated with JAK3 Activation.
Long-term Benefit of PD-L1 Blockade in Lung Cancer Associated with JAK3 Activation.
Cancer immunology research (2015-05-28)
Eliezer M Van Allen, Hadrien G Golay, Yan Liu, Shohei Koyama, Karrie Wong, Amaro Taylor-Weiner, Marios Giannakis, Maegan Harden, Vanesa Rojas-Rudilla, Aaron Chevalier, Tran Thai, Christine Lydon, Stacy Mach, Ada G Avila, Joshua A Wong, Alexandra R Rabin, Joshua Helmkamp, Lynette Sholl, Scott L Carter, Geoffrey Oxnard, Pasi Janne, Gad Getz, Neal Lindeman, Peter S Hammerman, Levi A Garraway, F Stephen Hodi, Scott J Rodig, Glenn Dranoff, Kwok-Kin Wong, David A Barbie
ABSTRACT
PD-1 immune checkpoint blockade occasionally results in durable clinical responses in advanced metastatic cancers. However, mechanism-based predictors of response to this immunotherapy remain incompletely characterized. We performed comprehensive genomic profiling on a tumor and germline sample from a patient with refractory lung adenocarcinoma who achieved marked long-term clinical benefit from anti-PD-L1 therapy. We discovered activating somatic and germline amino acid variants in JAK3 that promoted PD-L1 induction in lung cancer cells and in the tumor immune microenvironment. These findings suggest that genomic alterations that deregulate cytokine receptor signal transduction could contribute to PD-L1 activation and engagement of the PD-1 immune checkpoint in lung cancer.
Sigma-Aldrich
EGF from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
EGF from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Epidermal Growth Factor, human, animal component free, EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)
Sigma-Aldrich
EGF from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture