Merck
A new flavonoid glucoside from Rhododendron seniavinii.
Journal of Asian natural products research (2014-12-11)
Qing-Qing Wang, Chun Wu, Ying Zhang, Bai-Lian Liu, Guang-Xiong Zhou
ABSTRACT
The leaves of Rhododendron seniavinii Maxim with little phytochemical information are used as folk remedies for the treatment of acute and chronic bronchitis in China. In our pursuing for the biologically active chemical constituents in the leaves, a new flavonoid glycoside 5,7,3'-trimethoxy-quercetin-3-O-β-D-glucopyranoside (1) was isolated from the water extract of its leaves, together with two known compounds 5,7,3'-trimethoxy-quercetin (2) and ovafolinin B-9'-O-β-D-glucopyranoside (3). The structures of the new flavonoid glucoside as well as two known compounds were elucidated by spectroscopic and chemical methods.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications