Merck
  • Influence of Melatonin on the Proliferative and Apoptotic Responses of the Prostate under Normal and Hyperglycemic Conditions.
Influence of Melatonin on the Proliferative and Apoptotic Responses of the Prostate under Normal and Hyperglycemic Conditions.
Journal of diabetes research (2015-08-22)
Marina G Gobbo, Nishtman Dizeyi, Per-Anders Abrahamsson, Per-Anders Bertilsson, Viviane Sanches Masitéli, Eloisa Zanin Pytlowanciv, Sebastião R Taboga, Rejane M Góes
ABSTRACT
The antitumor properties of melatonin (MLT) are known for prostate cancer cells. This study investigated whether MLT affects prostate maturation and interferes with tissue injuries induced by diabetes. MLT was administered to Wistar rats from 5 weeks of age in the drinking water (10 μg/kg b.w.), and diabetes was induced at the 13th week by streptozotocin (4.5 mg/100g b.w., i.p.). The animals were euthanized in the 14th and 21st weeks. MLT reduced the immunostained cells for androgen receptor (AR) by 10% in younger rats. Diabetes decreased cell proliferation and increased apoptosis. MLT treatment impeded apoptosis (p = 0.02) and augmented proliferation (p = 0.0008) and PCNA content in prostate following long-term diabetes due to restoration of testosterone levels and expression of melatonin receptor type 1B. The effect of MLT (500 µM, 5 mM, and 10 mM) on androgen-dependent (22Rv1) and androgen-independent (PC3) cancer cells and human prostate epithelial cells (PNTA1) under normal and hyperglycemic conditions (HG, 450 mg/dL) was analyzed. Contrary to PNTA1 and 22Rv1 cells, MLT improved the proliferation of PC3 cells in hyperglycemic medium. The combined data indicated that MLT had proliferative and antiapoptotic effects in prostate cells subjected to HG levels and it seems to involve specific MLT pathways rather than AR.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Pricing and availability is not currently available.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Pricing and availability is not currently available.
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Pricing and availability is not currently available.
SAFC
Sodium chloride solution, 5 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, tablet
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Pricing and availability is not currently available.