Merck
  • Collaborative studies on the development of national reference standards for potency determination of H7N9 influenza vaccine.
Collaborative studies on the development of national reference standards for potency determination of H7N9 influenza vaccine.
Human vaccines & immunotherapeutics (2015-05-15)
Changgui Li, Kangwei Xu, Anwar Hashem, Ming Shao, Shuzhen Liu, Yong Zou, Qiang Gao, Yongchao Zhang, Liyong Yuan, Miao Xu, Xuguang Li, Junzhi Wang
ABSTRACT
The outbreak of human infections of a novel avian influenza virus A (H7N9) prompted the development of the vaccines against this virus. Like all types of influenza vaccines, H7N9 vaccine must be tested for its potency prior to being used in humans. However, the unavailability of international reference reagents for the potency determination of H7N9 vaccines substantially hinders the progress in vaccine development. To facilitate clinical development, we enlisted 5 participants in a collaborative study to develop critical reagents used in Single Radial Immunodiffusion (SRID), the currently acceptable assay for potency determination of influenza vaccine. Specifically, the hemagglutinin (HA) content of one vaccine bulk for influenza A (H7N9), herein designated as Primary Liquid Standard (PLS), was determined by SDS-PAGE. In addition, the freeze-dried antigen references derived from PLS were prepared to enhance the stability for long term storage. The final HA content of lyophilized antigen references were calibrated against PLS by SRID assay in a collaborative study. Importantly, application of these national reference standards to potency analyses greatly facilitated the development of H7N9 vaccines in China.
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 g
In Stock
Details...
€34.30
250 g
In Stock
Details...
€89.60
1 kg
In Stock
Details...
€271.00
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 g
In Stock
Details...
€34.30
250 g
In Stock
Details...
€89.60
1 kg
In Stock
Details...
€271.00
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 g
In Stock
Details...
€34.30
250 g
In Stock
Details...
€89.60
1 kg
In Stock
Details...
€271.00
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 g
In Stock
Details...
€34.30
250 g
In Stock
Details...
€89.60
1 kg
In Stock
Details...
€271.00
SAFC
MOPS
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 g
In Stock
Details...
€34.30
250 g
In Stock
Details...
€89.60
1 kg
In Stock
Details...
€271.00