Merck
  • Thio-glucose bound gold nanoparticles enhance radio-cytotoxic targeting of ovarian cancer.
Thio-glucose bound gold nanoparticles enhance radio-cytotoxic targeting of ovarian cancer.
Nanotechnology (2011-06-10)
Feng Geng, Kun Song, James Z Xing, Cunzhong Yuan, Shi Yan, Qifeng Yang, Jie Chen, Beihua Kong
ABSTRACT
The treatment of ovarian cancer has traditionally been intractable, and required novel approaches to improve therapeutic efficiency. This paper reports that thio-glucose bound gold nanoparticles (Glu-GNPs) can be used as a sensitizer to enhance ovarian cancer radiotherapy. The human ovarian cancer cells, SK-OV-3, were treated by gold nanoparticles (GNPs) alone, irradiation alone, or GNPs in addition to irradiation. Cell uptake was assayed using inductively coupled plasma atomic emission spectroscopy (ICP-AES), while cytotoxicity induced by radiotherapy was measured using both 3-(4,5)-dimethylthiahiazo (-z-y1)-3,5-di-phenytetrazoliumromide and clonogenic assays. The presence of reactive oxygen species (ROS) was determined using CM-H2-DCFDA confocal microscopy and cell apoptosis was determined by an Annexin V-FITC/propidium iodide (PI) kit with flow cytometry. The cells treated by Glu-GNPs resulted in an approximate 31% increase in nanoparticle uptake compared to naked GNPs (p < 0.005). Compared to the irradiation alone treatment, the intracellular uptake of Glu-GNPs resulted in increased inhibition of cell proliferation by 30.48% for 90 kVp and 26.88% for 6 MV irradiation. The interaction of x-ray radiation with GNPs induced elevated levels of ROS production, which is one of the mechanisms by which GNPs can enhance radiotherapy on ovarian cancer.
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Pricing and availability is not currently available.