Merck
  • Molecular cloning, tissue distribution, and chromosomal mapping of the human epithelial Ca2+ channel (ECAC1).
Molecular cloning, tissue distribution, and chromosomal mapping of the human epithelial Ca2+ channel (ECAC1).
Genomics (2000-08-17)
D Müller, J G Hoenderop, I C Meij, L P van den Heuvel, N V Knoers, A I den Hollander, P Eggert, V García-Nieto, F Claverie-Martín, R J Bindels
ABSTRACT
Functional and morphological analyses indicated that the epithelial Ca2+ channel (ECaC), which was recently cloned from rabbit kidney, exhibits the defining properties for being the gatekeeper in transcellular Ca2+ (re)absorption. Its human homologue provides, therefore, a molecular basis for achieving a better understanding of Ca2+ mal(re)absorption. By applying the RACE technique, the full-length cDNA of human ECaC (HGMW-approved symbol ECAC1) was obtained. It consisted of 2,772 bp with an open reading frame of 2,187 bp encoding a protein of 729 amino acids with a predicted molecular mass of 83 kDa. Phylogenetic analysis indicated that this highly selective Ca2+ channel exhibits a low level of homology (<30%) to other Ca2+ channels, suggesting that it belongs to a new family. hECaC was highly expressed in kidney, small intestine, and pancreas, and less intense expression was detected in testis, prostate, placenta, brain, colon, and rectum. These ECaC-positive tissues also expressed the 1,25-dihydroxyvitamin D3-sensitive calcium-binding proteins, calbindin-D9K and/or calbindin-D28K. The human ECaC gene mapped to chromosome 7q31.1-q31.2. Taken together, the conspicuous colocalization of hECaC and calbindins in organs that are not prime regulators of plasma Ca2+ levels could illustrate new pathways in cellular Ca2+ homeostasis.