Merck
  • Design, synthesis and biological evaluation of novel peptide MC2 analogues from Momordica charantia as potential anti-diabetic agents.
Design, synthesis and biological evaluation of novel peptide MC2 analogues from Momordica charantia as potential anti-diabetic agents.
Organic & biomolecular chemistry (2015-03-18)
Baowei Yang, Xue Li, Chenyu Zhang, Sijia Yan, Wei Wei, Xuekun Wang, Xin Deng, Hai Qian, Haiyan Lin, Wenlong Huang
ABSTRACT
Three series of Momordica charantia (MC)2 analogues were designed, synthesized and evaluated for their anti-hyperglycaemic effects. Alanine scanning focusing on the peptide MC2 indicated the importance of the residues proline (Pro)(3), serine (Ser)(6), isoleucine (Ile)(7) and Ser(10) for anti-hyperglycaemic effects. Among the first series of MC2 analogues, peptide I-4 exhibited a better anti-hyperglycaemic effect and was chosen for further modification. A further two series of conformationally constrained analogues were designed by scanning the residues Pro(3), Ser(6), Ile(7), and Ser(10) with an i - (i + 2) lactam bridge consisting of a glutamic acid-Xaa-lysine (Glu-Xaa-Lys) scaffold and a diproline fragment. By screening in normal mice and mice with diabetes mellitus, peptides II-1, II-2 and III-3 showed a significant improvement in anti-hyperglycaemic and anti-oxidative activities compared with I-4. These data suggest that II-1, II-2 and III-3 could be candidates for future treatment of diabetes mellitus.
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, for molecular biology, 1 M in H2O
Sigma-Aldrich
Citric acid trisodium salt, ≥98% (GC), anhydrous
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
N-Ethyldiisopropylamine, BASF quality, ≥98.0%
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
Supelco
Sodium Citrate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
Sigma-Aldrich
HBTU, ≥98.0% (T)
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%
Sigma-Aldrich
N,N-Diisopropylethylamine, 99.5%, biotech. grade