Merck
  • Reduced plasma levels of soluble interleukin-7 receptor during graft-versus-host disease (GVHD) in children and adults.
Reduced plasma levels of soluble interleukin-7 receptor during graft-versus-host disease (GVHD) in children and adults.
BMC immunology (2014-06-21)
Thomas Poiret, Lalit Rane, Mats Remberger, Birgitta Omazic, Asa Gustafsson-Jernberg, Nalini Kumar Vudattu, Raija Ahmed, Ingemar Ernberg, Jacek Winiarski, Isabelle Magalhaes, Olle Ringden, Markus Maeurer
ABSTRACT
Interleukin 7 (IL-7) signals via the IL-7 receptor (IL-7R) and drives homeostatic T-cell proliferation in patients after allogeneic hematopoietic stem cell transplantation (aHSCT). We performed a prospective study in adults (n = 33) and children (n = 29) undergoing aHSCT measuring plasma IL-7 and soluble IL-7R (sIL-7R) concentrations between 1 and 12 months after HSCT in order to investigate the link between sIL-7R and clinical events after aHSCT. sIL-7R, but not IL-7, increased with time after HSCT in plasma from all patients enrolled in the study. sIL-7R values were higher at 2, 3, and 6 months (p < 0.01) if the donor was a sibling as compared to an unrelated donor. Increased sIL-7R levels were also identified in plasma from patients who were not treated with anti-thymocyte globulin (ATG). Low sIL-7R was associated with any grade of acute graft-versus-host disease (GVHD) at 2 and 6 months (p = 0.02) and with a positive CMV PCR at 2 months after HSCT (p < 0.05). Patients with cytomegalovirus (CMV) reactivation had increased IL-7 values at 2 and 3 months (p = 0.02) after HSCT. In multivariate analysis, lower sIL-7R levels were associated with acute GVHD (relative hazard (RH): 0.70, p > 0.01) and sibling donors (RH: 2.23, p = 0.004). Recipients of sibling grafts showed high levels of IL-7 (RH: 1.38, p < 0.05) and bone marrow recipients had low IL-7 levels (RH: 0.73, p = 0.04). Measurement of the sIL-7R/IL-7 axis will help in guided immune monitoring after HSCT and guided interference with sIL-7R may be explored in GVHD management.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Interleukin-7 human, ≥98% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
IL-7 human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)