Merck
  • Astrocytic JWA expression is essential to dopaminergic neuron survival in the pathogenesis of Parkinson's disease.
Astrocytic JWA expression is essential to dopaminergic neuron survival in the pathogenesis of Parkinson's disease.
CNS neuroscience & therapeutics (2014-03-19)
Shu-Han Miao, Hong-Bin Sun, Yang Ye, Jing-Jing Yang, Yao-Wei Shi, Ming Lu, Gang Hu, Jian-Wei Zhou
ABSTRACT
To investigate the role of astrocytic JWA expression in dopaminergic (DA) neuron degeneration and in the pathogenesis of Parkinson's disease (PD). Conditional astrocytic JWA null (JWA∆2/∆2/GFAP-Cre) mice and U251 glioma cells were used to evaluate the effects of JWA gene on DA neuron degeneration. The oxidative stress-driven molecular events were determined in both in vivo and in vitro models. Conditional astrocytic JWA knockout resulted in significant activation of astrocytes measured by increase in glial fibrillary acidic protein-positive cells (1.34×10(3)±74.5 vs. 8.44×10(3)±1.35×10(3), P<0.01) in mouse substantia nigra, accompanied by loss of DA neurons (1.03×10(4)±238 vs. 6.17×10(3)±392, P<0.001). Deficiency of JWA significantly aggravated reactive oxygen species (ROS) accumulation in substantia nigra compared with the wild-type mice. Increasing JWA expression in U251 glioma cells inhibited ROS with a concomitant increase in intracellular glutathione. Furthermore, suppression of IKKβ-nuclear factor (NF)-κB signaling pathway was shown to regulate JWA in a PD model. The JWA gene exerts neuroprotective roles against DA neuronal degeneration via modulating intracellular redox status and NF-κB signaling pathway and is a potential treatment target for PD.
Sigma-Aldrich
Anti-LCP1 (ab1) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Dihydroethidium, BioReagent, suitable for fluorescence, ≥95% (HPCE)
Sigma-Aldrich
Dihydroethidium, ≥95%
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder