Merck
  • The protective effects of glutamine in a rat model of ventilator-induced lung injury.
The protective effects of glutamine in a rat model of ventilator-induced lung injury.
Journal of thoracic disease (2015-01-16)
Chin-Ming Chen, Kuo-Chen Cheng, Chien-Feng Li, Haibo Zhang
ABSTRACT
The mortality rate of patients with acute respiratory distress syndrome (ARDS) is still high despite the use of protective ventilatory strategies. We sought to examine the pharmacological effects of glutamine (GLN) in a two-hit model of endotoxin-induced inflammation followed by ventilator-induced lung injury (VILI). We hypothesized that the administration of GLN ameliorates the VILI. Sprague-Dawley rats were anesthetized and given lipopolysaccharide (LPS) intratracheally as a first hit to induce lung inflammation, followed 24 h later by a second hit of mechanical ventilation (MV) with either low tidal volume (6 mL/kg) with 5 cmH2O of positive end-expiratory pressure (PEEP) or high tidal volume (22 mL/kg) with zero PEEP for 4 h. GLN or lactated Ringer's solution as the placebo was administered intravenously 15 min prior to MV. In the LPS-challenged rats ventilated with high tidal volume, the treatment with GLN improved lung injury indices, lung mechanics and cytokine responses compared with the placebo group. The administration of GLN given immediately prior to MV may be beneficial in the context of reducing VILI.
Sigma-Aldrich
Urethane, ≥99%
Sigma-Aldrich
Urethane, ≥99.0% (GC)
Sigma-Aldrich
2,3-Dimercapto-1-propanol, ≥98% (iodometric)
Supelco
Urethane, analytical standard