Merck
  • Physicochemical and drug metabolism characterization of a series of 4-aminoquinoline-3-hydroxypyridin-4-one hybrid molecules with antimalarial activity.
Physicochemical and drug metabolism characterization of a series of 4-aminoquinoline-3-hydroxypyridin-4-one hybrid molecules with antimalarial activity.
Expert opinion on drug metabolism & toxicology (2014-09-16)
Roslyn Thelingwani, Britta Bonn, Kelly Chibale, Collen Masimirembwa
ABSTRACT
Drug resistance by Plasmodium falciparum remains a challenge in malaria chemotherapy. This paper will focus on physicochemical and drug metabolism characterization of a series of 4-aminoquinoline-3-hydroxypyridin-4-one hybrid shown to have antimalarial activity against chloroquine-resistant P. falciparum. The aim is to identify potential liabilities that could arise from covalently linking two pharmacophoric units of different drug classes into one functional drug. Metabolism and biotransformation of hybrids were investigated using cryopreserved human hepatocytes. Generic incubation conditions with respect to substrate concentration, cell concentration and incubation time were used. Evaluation of the hybrids for metabolism-based drug-drug interaction (DDI) risk was done with recombinant CYPs. A generic UPLC-MSMS bioanalytical method was used for quantitation and metabolite identification. Prediction of absorption, distribution, metabolism, excretion and toxicity (ADMET) properties of the hybrid compounds was done using various software. The analogs generally had good physicochemical profiles. Metabolism was mainly via the linker chain and was predicted to be intermediate to fast in cryopreserved human hepatocytes. However, analogs had potential to cause DDIs as they were potent inhibitors of CYP3A4 and CYP2D6. Although the efficacy of the compounds is promising, they carry a potentially limiting liability of CYP inhibition and rapid metabolic clearance.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Supelco
Erythromycin, Pharmaceutical Secondary Standard; Certified Reference Material
Erythromycin, for microbiological assay, European Pharmacopoeia (EP) Reference Standard
USP
Erythromycin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Erythromycin standard solution, 1 mg/mL in H2O
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
7-Methoxy-4-(trifluoromethyl)coumarin, ≥99% (TLC)
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Ketoconazole