Merck
  • Development of a community-based intervention for the control of Chagas disease based on peridomestic animal management: an eco-bio-social perspective.
Development of a community-based intervention for the control of Chagas disease based on peridomestic animal management: an eco-bio-social perspective.
Transactions of the Royal Society of Tropical Medicine and Hygiene (2015-01-22)
Sandra M De Urioste-Stone, Pamela M Pennington, Elizabeth Pellecer, Teresa M Aguilar, Gabriela Samayoa, Hugo D Perdomo, Hugo Enríquez, José G Juárez
ABSTRACT
Integrated vector management strategies depend on local eco-bio-social conditions, community participation, political will and inter-sectorial partnership. Previously identified risk factors for persistent Triatoma dimidiata infestation include the presence of rodents and chickens, tiled roofs, dirt floors, partial wall plastering and dog density. A community-based intervention was developed and implemented based on cyclical stakeholder and situational analyses. Intervention implementation and evaluation combined participatory action research and cluster randomized pre-test post-test experimental designs. The intervention included modified insecticide application, education regarding Chagas disease and risk factors, and participatory rodent control. At final evaluation there was no significant difference in post-test triatomine infestation between intervention and control, keeping pre-test rodent and triatomine infestations constant. Knowledge levels regarding Chagas disease and prevention practices including rodent control, chicken management and health service access increased significantly only in intervention communities. The odds of nymph infection and rat infestation were 8.3 and 1.9-fold higher in control compared to intervention communities, respectively. Vector control measures without reservoir control are insufficient to reduce transmission risk in areas with persistent triatomine infestation. This integrated vector management program can complement house improvement initiatives by prioritizing households with risk factors such as tiled roofs. Requirement for active participation and multi-sectorial coordination poses implementation challenges.
Product Description
Supelco
Deltamethrin, PESTANAL®, analytical standard
Supelco
Deltamethrin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland