Merck
  • Long-term outcomes of needle revision of failing deep sclerectomy blebs.
Long-term outcomes of needle revision of failing deep sclerectomy blebs.
Graefe's archive for clinical and experimental ophthalmology = Albrecht von Graefes Archiv fur klinische und experimentelle Ophthalmologie (2014-10-12)
Antigoni Koukkoulli, Fayyaz Musa, Nitin Anand
ABSTRACT
To report on long-term outcomes of mitomycin C (MMC)-augmented needle revision of failing deep sclerectomy (DS) blebs. Retrospective database search of all needle revisions with MMC for DS blebs between 2002 and 2008 was conducted. Sixty-six eyes of 66 patients were included in the study. Subconjunctival MMC 0.01-0.02 mg was injected 15 min before needle revision. Complete success was defined as intraocular pressure ≤ 18 mmHg or 20% decrease from baseline with no glaucoma medications or further surgical procedures. Partial success was based on the same criteria, but with medications. Mean follow-up after index needle revision was 67.8 ± 24.8 months (range 1-10 years). The mean number of needle revisions was 1.6 ± 0.8 (range 1-4). Two or more procedures were done in 31 eyes (47.0%). Mean preoperative intraocular pressure was 23.2 ± 6.9 mmHg. Complete and partial success rates were 64% and 71% at 1 year, 57% and 68% at 3 years, and 40 % and 58 % at 5 years after surgery, respectively. Early needle revision, within 6 months of index surgery, was significantly associated with failure, both on uni- and multivariate tests. Significant complications included delayed hypotony in five eyes (7.6%), delayed bleb leak in two eyes (3.0%), endophthalmitis in one eye (1.5%), and corneal oedema in two eyes (3.0%). Needle revision with subconjunctival MMC may successfully lower the intraocular pressure in eyes with a failing DS bleb in the long term. Complications known to be associated with trabeculectomy and MMC were anticipated and observed in our DS group.
Product Description
USP
Prednisolone acetate, United States Pharmacopeia (USP) Reference Standard
Prednisolone acetate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Prednisolone 21-acetate, ≥97%
Prednisolone acetate for peak identification, European Pharmacopoeia (EP) Reference Standard