Merck
  • Mandibular advancement device and CPAP upon cardiovascular parameters in OSA.
Mandibular advancement device and CPAP upon cardiovascular parameters in OSA.
Sleep & breathing = Schlaf & Atmung (2014-01-28)
Cibele Dal-Fabbro, Silvério Garbuio, Vânia D'Almeida, Fátima D Cintra, Sergio Tufik, Lia Bittencourt
ABSTRACT
This study aims to compare the effects of a mandibular advancement device (MAD) with continuous positive airway pressure (CPAP) treatment for obstructive sleep apnea (OSA) on blood pressure (BP), oxidative stress, and heart rate variability (HRV) in a randomized, crossed-over, single-blind, and controlled trial. Twenty-nine moderate-to-severe adult OSA patients underwent MAD, CPAP, and placebo oral appliance treatment. Polysomnography, Epworth sleepiness scale, 24-h ambulatory BP monitoring, oxidative stress parameters (malondialdehyde, catalase, superoxide dismutase, vitamins C, E, B6, B12, folate, homocysteine, uric acid), and HRV were assessed at baseline and after 1 month of each treatment. Diaries were used to evaluate compliance for devices and a pressure-time meter for CPAP. Both active treatments resulted in decreases in apnea and hypopnea index and Epworth sleepiness scale; CPAP showed a greater effect. Frequency of diastolic BP dipping was higher in the MAD group compared with the CPAP group. A significant drop from baseline levels for catalase activity was observed after MAD. For HRV, there was a significant decrease in total power at night with CPAP and MAD compared with POA, and a decrease in index of sleep autonomic variation with MAD compared with baseline levels. Compliance rates were higher with MAD rather than CPAP. Even though CPAP proved to be more effective at attenuating OSA, better compliance with MAD favored the reduction of one of the enzymes which participates in oxidative stress and better autonomic modulation during sleep.
Sigma-Aldrich
Cyanocobalamin, tested according to Ph. Eur.
Pricing and availability is not currently available.
Supelco
Cyanocobalamin (B12), analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Pricing and availability is not currently available.
Cyanocobalamin, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Cyanocobalamin, British Pharmacopoeia (BP) Assay Standard
Pricing and availability is not currently available.
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material
Pricing and availability is not currently available.
Sigma-Aldrich
Vitamin B12, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Pricing and availability is not currently available.