Merck
  • An epigenetic marker panel for recurrence risk prediction of low grade papillary urothelial cell carcinoma (LGPUCC) and its potential use for surveillance after transurethral resection using urine.
An epigenetic marker panel for recurrence risk prediction of low grade papillary urothelial cell carcinoma (LGPUCC) and its potential use for surveillance after transurethral resection using urine.
Oncotarget (2014-07-02)
Leonel Maldonado, Mariana Brait, Christina Michailidi, Enrico Munari, Tina Driscoll, Luciana Schultz, Trinity Bivalacqua, Mark Schoenberg, David Sidransky, George J Netto, Mohammad Obaidul Hoque
ABSTRACT
By a candidate gene approach, we analyzed the promoter methylation (PM) of 8 genes genes (ARF, TIMP3, RAR-β2, NID2, CCNA1, AIM1, CALCA and CCND2) by quantitative methylation specific PCR (QMSP) in DNA of 17 non-recurrent and 19 recurrent noninvasive low grade papillary urothelial cell carcinoma (LGPUCC) archival tissues. Among the genes tested, by establishing an empiric cutoff value, CCND2, CCNA1, NID2, and CALCA showed higher frequency of methylation in recurrent than in non-recurrent LGPUCC: CCND2 10/19 (53%) vs. 2/17 (12%) (p=0.014); CCNA1 11/19 (58%) vs. 4/17 (23.5%) (p=0.048); NID2 13/19 (68%) vs. 3/17 (18%) (p=0.003) and CALCA 10/19 (53%) vs. 4/17 (23.5%) (p=0.097), respectively. We further analyzed PM of CCND2, CCNA1, and CALCA in urine DNA from UCC patients including LGPUCC and controls. The frequency of CCND2, CCNA1 and CALCA was significantly higher (p<0.0001) in urine of UCC cases [ 38/148 (26%), 50/73 (68%) and 94/148 (63.5%) respectively] than controls [0/56 (0%), 10/60 (17%) and 16/56 (28.5%), respectively)]. Most importantly we found any one of the 3 markers methylation positive in 25 out of 30 (83%) cytology negative LGPUCC cases. We also explored the biological function of CCNA1 in UCC. Prospective confirmatory studies are needed to develop a reliable tool for prediction of recurrence using primary LGPUCC tissues and/or urine.
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard