Merck
  • Krüppel-like factor 8 involved in hypoxia promotes the invasion and metastasis of gastric cancer via epithelial to mesenchymal transition.
Krüppel-like factor 8 involved in hypoxia promotes the invasion and metastasis of gastric cancer via epithelial to mesenchymal transition.
Oncology reports (2014-10-22)
Na Liu, Yafang Wang, Yongan Zhou, Hailin Pang, Jing Zhou, Pei Qian, Lili Liu, Helong Zhang
ABSTRACT
Previously, we reported that hypoxia was able to induce invasion and metastasis in gastric cancer and that hypoxia-inducible factor-1 (HIF-1) is a key factor involved in this tumor type. Krüppel-like factor 8 (KLF8) as a transcriptional repressor has been suggested as a promoter of tumor metastasis in breast cancer and an inducer of the epithelial‑mesenchymal transition (EMT). KLF8 is also highly expressed in gastric cancer tissues, contributing to poor prognosis. However, the association between KLF8 and HIF-1 in regulating the progression of human gastric cancer in hypoxia is unclear. In the present study, we found that KLF8 was overexpressed in gastric cancer metastatic tissues and cells. Additionally, KLF8 siRNA significantly inhibited SGC7901 cell invasion and migration compared with SGC7901, SGC7901/Scr-si cells. Hypoxia is thus able to induce KLF8 expression and EMT in SGC7901 cells. However, following the examination of changes in cell morphology and epithelial and mesenchymal markers, it was found that KLF8 siRNA and HIF-1 siRNA strongly reversed EMT in cells undergoing hypoxia. Furthermore, hypoxia-induced KLF8 overexpression was attenuated by HIF-1 siRNA. Experiments using luciferase promoter constructs resulted in a marked increase in the activity of cells exposed to hypoxia and decreased activity in cells co-transfected with HIF-1 siRNA. The chromatin immunoprecipitation assay revealed proximal HRE at -133 is the main HIF-1 binding site in the KLF8 promoter. In conclusion, the results demonstrated that KLF8 is actively enhanced by hypoxia and is a novel HIF-1 target. KLF8 is a novel EMT regulating transcription factor that involved in the progression of gastric cancer. The specific anti-EMT drugs in combination with anti-hypoxia are new promising cancer therapies.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Setd2
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
MISSION® esiRNA, targeting human SETD2
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Kif9