Merck
  • Neurocognitive performance and serial intravenous subanesthetic ketamine in treatment-resistant depression.
Neurocognitive performance and serial intravenous subanesthetic ketamine in treatment-resistant depression.
The international journal of neuropsychopharmacology (2014-06-26)
Paulo R Shiroma, C Sophia Albott, Brian Johns, Paul Thuras, Joseph Wels, Kelvin O Lim
ABSTRACT
The N-methyl-D-aspartate glutamate receptor antagonist ketamine has demonstrated rapid antidepressant effects in treatment-resistant depression (TRD). However, evaluation of ketamine's neurocognitive aspects in TRD has started to be explored. This study aims to (1) examine baseline neurocognitive performance and change in severity of depressive symptoms through six ketamine infusions, (2) examine the neurocognitive effects after completion of serial infusions and whether changes were associated to relapse to depression. Six IV infusions of 0.5 mg/Kg ketamine over 40 min were conducted on a Monday-Wednesday-Friday schedule during a 12-d period on 15 patients with TRD followed by a 4-wk observational period. Neurocognitive functioning was assessed using the CogState battery at baseline and at each follow-up visit. Tasks were designed to test attention, memory (working, visual, and verbal), speed of processing, and set shifting. The likelihood of response through six infusions was greater among depressed subjects with lower attention at baseline (F(1,13)=5.59, p=0.034). Significant improvement was found in scores of visual memory (F(4,33.82)=5.12, p=0.002), simple working memory (F(4, 24.85)=3.29, p=0.027) and complex working memory (F(4, 32.76)=4.18, p=0.008) after the last ketamine infusion. However, neurocognitive changes were accounted for by improvement in the severity of depressive symptom. The acute neurocognitive effect after completion of repeated infusions was not associated with the likelihood of subsequent relapse during follow-up. Our findings suggest a potential baseline neurocognitive predictor of ketamine response and the apparently lack of short-term neurocognitive impairment after completion of six ketamine infusions in TRD.
Product Description
Ketamine hydrochloride, European Pharmacopoeia (EP) Reference Standard