Merck
  • Evaluation of the relationship between polymorphisms in CYP2C19 and the pharmacokinetics of omeprazole, pantoprazole and rabeprazole.
Evaluation of the relationship between polymorphisms in CYP2C19 and the pharmacokinetics of omeprazole, pantoprazole and rabeprazole.
Pharmacogenomics (2014-12-17)
Manuel Román, Dolores Ochoa, Sergio Daniel Sánchez-Rojas, Maria Talegón, Rocio Prieto-Pérez, Ângela Rivas, Francisco Abad-Santos, Teresa Cabaleiro
ABSTRACT
To evaluate the possible association between polymorphisms in CYP2C19 and the pharmacokinetics of omeprazole, rabeprazole and pantoprazole. 151 healthy volunteers were evaluated for polymorphisms in the CYP2C19 gene using real-time polymerase chain reaction. Plasma concentrations were measured using high-performance liquid chromatography coupled to mass spectrometry. Carriers of the *2 allele displayed poor metabolism for all the PPIs studied (around 50% decrease in clearance). Subjects with the *17 allele showed a light increase in clearance compared with *1/*1 (not significant). CYP2C19*2 is associated with decreased clearance of all the PPIs, that could be associated with higher drug efficacy. CYP2C19*17 could increase clearance of these drugs, although the effect seems small.