Merck
  • Construction of carotenoid biosynthetic pathways using squalene synthase.
Construction of carotenoid biosynthetic pathways using squalene synthase.
FEBS letters (2013-12-18)
Maiko Furubayashi, Ling Li, Akinori Katabami, Kyoichi Saito, Daisuke Umeno
ABSTRACT
The first committed steps of steroid/hopanoid pathways involve squalene synthase (SQS). Here, we report the Escherichia coli production of diaponeurosporene and diapolycopene, yellow C30 carotenoid pigments, by expressing human SQS and Staphylococcus aureus dehydrosqualene (C30 carotenoid) desaturase (CrtN). We suggest that the carotenoid pigments are synthesized mainly via the desaturation of squalene rather than the direct synthesis of dehydrosqualene through the non-reductive condensation of prenyl diphosphate precursors, indicating the possible existence of a "squalene route" and a "lycopersene route" for C30 and C40 carotenoids, respectively. Additionally, this finding yields a new method of colorimetric screening for the cellular activity of squalene synthases, which are major targets for cholesterol-lowering drugs.